Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rig-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001375088.2 | NP_001362001.1 | 17 - 18 | 3144 | 71 | |
NM_001383616.1 | 16 - 17 | 3099 | 71 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |