Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: vav-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001047757.3 | NP_001041222.1 | 16 - 17 | 2841 | 66 | |
NM_001373412.3 | NP_001359918.1 | 17 - 18 | 2997 | 66 | ||
NM_001373413.3 | NP_001360680.1 | 17 - 18 | 2984 | 66 | ||
GenBank mRNA | DQ136172.1 | - | 17 - 18 | 2935 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |