Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gap-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001307914.4 | NP_001294843.1 | 2 - 3 | 234 | 104 | |
NM_001307916.4 | NP_001294845.1 | 7 - 8 | 1011 | 104 | ||
NM_001383623.1 | 5 - 6 | 615 | 104 | |||
NM_077193.6 | NP_509594.3 | 7 - 8 | 1012 | 104 | ||
NM_077194.7 | NP_509595.1 | 8 - 9 | 1074 | 104 | ||
NM_077195.7 | NP_509596.1 | 6 - 7 | 1662 | 104 | ||
NM_077196.6 | NP_509597.1 | 5 - 6 | 801 | 104 | ||
NM_077197.6 | NP_509598.1 | 4 - 5 | 551 | 104 | ||
NM_077198.7 | NP_509599.1 | 2 - 3 | 1131 | 104 | ||
NM_171753.7 | NP_741873.1 | 5 - 6 | 1034 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 Caenorhabditis elegans Probe spans exons |