Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: acl-5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001047817.4 | NP_001041282.1 | 3 - 4 | 352 | 91 | |
NM_001047818.4 | NP_001041283.1 | 2 - 3 | 180 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |