Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cut-5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001270170.4 | NP_001257099.1 | 9 - 10 | 1008 | 73 | |
NM_001270171.3 | NP_001257100.1 | 8 - 9 | 1045 | 73 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |