Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mig-15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029800.3 | NP_001024971.1 | 1 - 2 | 122 | 73 | |
NM_001029801.5 | NP_001024972.1 | 1 - 2 | 122 | 73 | ||
NM_001029802.5 | NP_001024973.1 | 1 - 2 | 46 | 73 | ||
NM_001029803.6 | NP_001024974.1 | 1 - 2 | 129 | 73 | ||
GenBank mRNA | AF087131.1 | - | 1 - 2 | 144 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |