Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lgc-41 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001129644.2 | NP_001123116.1 | 7 - 8 | 804 | 120 | |
NM_001373473.2 | NP_001360683.1 | 7 - 8 | 804 | 120 | ||
NM_001373474.1 | NP_001359532.1 | 7 - 8 | 804 | 120 | ||
NM_001373475.2 | NP_001360682.1 | 7 - 8 | 805 | 120 | ||
NM_001373476.2 | NP_001360686.1 | 7 - 8 | 804 | 120 | ||
NM_001373477.1 | NP_001360687.1 | 7 - 8 | 804 | 120 | ||
NM_001373478.3 | NP_001360684.1 | 8 | 847 | 120 | ||
NM_001373479.2 | NP_001360685.1 | 8 - 9 | 852 | 120 | ||
NM_001373480.3 | NP_001360688.1 | 8 - 9 | 853 | 120 | ||
NM_001373481.2 | NP_001360689.1 | 8 | 843 | 120 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |