Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: daf-12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029376.6 | NP_001024547.1 | 11 - 12 | 1110 | 81 | |
NM_001047774.5 | NP_001041239.1 | 10 - 11 | 1008 | 81 | ||
NM_001381088.1 | NP_001366986.1 | 9 - 10 | 933 | 81 | ||
GenBank mRNA | AF136238.1 | - | 11 - 12 | 1143 | 81 | |
AF136239.1 | - | 9 - 10 | 940 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |