Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: daf-12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029376.6 | NP_001024547.1 | 2 - 3 | 174 | 74 | |
GenBank mRNA | AF136238.1 | - | 2 - 3 | 207 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Caenorhabditis elegans Probe spans exons |