Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: daf-12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029376.6 | NP_001024547.1 | 9 - 10 | 932 | 65 | |
NM_001047774.5 | NP_001041239.1 | 8 - 9 | 830 | 65 | ||
NM_001381088.1 | NP_001366986.1 | 7 - 8 | 755 | 65 | ||
GenBank mRNA | AF136238.1 | - | 9 - 10 | 965 | 65 | |
AF136239.1 | - | 7 - 8 | 762 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |