Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: egl-36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_077394.4 | NP_509795.1 | 10 - 11 | 1584 | 122 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |