Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: egl-36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_077394.4 | NP_509795.1 | 4 - 5 | 413 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |