Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: plc-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029446.3 | NP_001024617.1 | 24 - 25 | 4615 | 72 | |
NM_001029448.6 | NP_001024619.1 | 24 - 25 | 4577 | 72 | ||
NM_001129669.4 | NP_001123141.2 | 57 - 58 | 9455 | 72 | ||
NM_001136454.3 | NP_001129926.1 | 24 - 25 | 4607 | 72 | ||
NM_001136455.4 | NP_001129927.1 | 24 - 25 | 4799 | 72 | ||
NM_001373489.2 | NP_001360637.1 | 57 - 58 | 9434 | 72 | ||
NM_001373491.2 | NP_001360639.1 | 57 - 58 | 9464 | 72 | ||
NM_001373492.2 | NP_001360635.1 | 57 - 58 | 9485 | 72 | ||
NM_001373493.2 | NP_001360636.1 | 58 - 59 | 9605 | 72 | ||
NM_001373494.2 | NP_001359720.1 | 58 - 59 | 9626 | 72 | ||
NM_001373495.2 | NP_001360638.1 | 58 - 59 | 9635 | 72 | ||
NM_077404.6 | NP_509805.3 | 24 - 25 | 4577 | 72 | ||
GenBank mRNA | AF044576.1 | - | 24 - 25 | 4637 | 72 | |
AM992255.1 | - | 24 - 25 | 4607 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |