Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: plc-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001129669.4 | NP_001123141.2 | 22 - 23 | 2934 | 75 | |
NM_001373489.2 | NP_001360637.1 | 22 - 23 | 2934 | 75 | ||
NM_001373491.2 | NP_001360639.1 | 22 - 23 | 2964 | 75 | ||
NM_001373492.2 | NP_001360635.1 | 22 - 23 | 2964 | 75 | ||
NM_001373493.2 | NP_001360636.1 | 23 - 24 | 3105 | 75 | ||
NM_001373494.2 | NP_001359720.1 | 23 - 24 | 3105 | 75 | ||
NM_001373495.2 | NP_001360638.1 | 23 - 24 | 3135 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |