Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: plc-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001129669.4 | NP_001123141.2 | 23 - 24 | 3099 | 80 | |
NM_001373492.2 | NP_001360635.1 | 23 - 24 | 3129 | 80 | ||
NM_001373494.2 | NP_001359720.1 | 24 - 25 | 3270 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Caenorhabditis elegans Probe spans exons |