Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pgp-6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001047822.6 | NP_001041287.2 | 1 - 2 | 138 | 128 | |
NM_001270189.3 | NP_001257118.1 | 1 - 2 | 73 | 128 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |