Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: apt-9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_077452.6 | NP_509853.2 | 7 - 8 | 1181 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |