Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sulp-7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038479.2 | NP_001033568.1 | 3 - 4 | 237 | 106 | |
NM_001038480.2 | NP_001033569.1 | 3 - 4 | 297 | 106 | ||
NM_001038481.4 | NP_001033570.1 | 5 - 6 | 569 | 106 | ||
NM_001038482.4 | NP_001033571.1 | 5 - 6 | 569 | 106 | ||
GenBank mRNA | AY887913.1 | - | 3 - 4 | 285 | 106 | |
AY887914.1 | - | 3 - 4 | 342 | 106 | ||
AY887915.1 | - | 5 - 6 | 591 | 106 | ||
AY887916.1 | - | 5 - 6 | 591 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |