Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: inx-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_077484.5 | NP_509885.1 | 3 - 4 | 1086 | 103 | |
GenBank mRNA | AF304127.1 | - | 3 - 4 | 1080 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |