Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pgp-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001270213.3 | NP_001257142.1 | 9 - 10 | 2003 | 107 | |
NM_001270214.5 | NP_001257143.1 | 8 - 9 | 1921 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |