Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: slo-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029356.4 | NP_001024527.1 | 9 - 10 | 1665 | 99 | |
NM_001029357.5 | NP_001024528.1 | 9 - 10 | 1665 | 99 | ||
NM_001029358.5 | NP_001024529.1 | 9 - 10 | 1346 | 99 | ||
NM_001136452.4 | NP_001129924.1 | 9 - 10 | 1349 | 99 | ||
NM_001270215.3 | NP_001257144.1 | 11 - 12 | 1457 | 99 | ||
NM_001270216.3 | NP_001257145.1 | 10 - 11 | 1412 | 99 | ||
NM_001270217.3 | NP_001257146.1 | 10 - 11 | 1466 | 99 | ||
NM_001270218.3 | NP_001257147.1 | 10 - 11 | 1427 | 99 | ||
GenBank mRNA | AF173828.1 | - | 9 - 10 | 1557 | 99 | |
AF232770.1 | - | 9 - 10 | 1358 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |