Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: frm-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001351892.4 | NP_001338849.1 | 5 | 470 | 82 | |
NM_001375142.3 | NP_001361874.1 | 4 - 5 | 440 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Caenorhabditis elegans Probe spans exons |