Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: frm-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001351892.4 | NP_001338849.1 | 6 | 572 | 112 | |
NM_001375142.3 | NP_001361874.1 | 5 - 6 | 542 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |