Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ttx-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001270239.2 | NP_001257168.1 | 5 - 6 | 602 | 133 | |
NM_001270240.3 | NP_001257169.1 | 4 - 5 | 518 | 133 | ||
GenBank mRNA | U72211.1 | - | 5 - 6 | 602 | 133 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |