Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tsp-20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_077613.7 | NP_510014.1 | 3 - 4 | 563 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |