Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lgc-24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001129638.3 | NP_001123110.2 | 7 - 8 | 956 | 143 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |