Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cab-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_077635.6 | NP_510036.1 | 4 - 5 | 500 | 102 | |
GenBank mRNA | AF293976.1 | - | 2 - 3 | 313 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |