Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: srd-40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_077661.2 | NP_510062.1 | 2 - 3 | 435 | 172 |
Set Membership: |
Amplicon length greater than or equal to 101 G-protein coupled receptor Detect Genomic DNA Caenorhabditis elegans Probe spans exons |