Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nekl-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_077679.8 | NP_510080.2 | 2 - 3 | 263 | 134 | |
GenBank mRNA | DQ645955.1 | - | 2 - 3 | 279 | 134 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |