Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: myo-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_077691.5 | NP_510092.2 | 9 - 10 | 5221 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |