Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: atf-6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_077693.6 | NP_510094.1 | 7 - 8 | 1230 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |