Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nhr-17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_077714.5 | NP_510115.1 | 5 - 6 | 605 | 139 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |