Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pqn-18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001270274.4 | NP_001257203.1 | 3 - 4 | 408 | 71 | |
NM_001270275.4 | NP_001257204.1 | 2 - 3 | 321 | 71 | ||
NM_001381127.1 | NP_001366740.1 | 5 - 6 | 1028 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |