Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ptr-24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001270290.5 | NP_001257219.1 | 4 - 5 | 931 | 96 | |
NM_001270291.2 | NP_001257220.1 | 3 - 4 | 709 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |