Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mxl-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_077822.6 | NP_510223.1 | 3 - 4 | 486 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |