Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: flr-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_077842.4 | NP_510243.1 | 14 - 15 | 1703 | 99 | |
GenBank mRNA | AB012617.1 | - | 14 - 15 | 1609 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |