Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tre-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_077848.8 | NP_510249.2 | 4 - 5 | 695 | 69 | |
GenBank mRNA | AJ512338.1 | - | 4 - 5 | 695 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |