Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: acr-12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001313441.4 | NP_001300370.1 | 4 - 5 | 514 | 95 | |
NM_077861.6 | NP_510262.1 | 6 - 7 | 977 | 95 | ||
GenBank mRNA | AF187013.1 | - | 6 - 7 | 928 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |