Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: acr-12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001313441.4 | NP_001300370.1 | 7 - 8 | 947 | 61 | |
NM_077861.6 | NP_510262.1 | 9 - 10 | 1410 | 61 | ||
GenBank mRNA | AF187013.1 | - | 9 - 10 | 1361 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |