Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: acr-9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_077884.5 | NP_510285.2 | 5 - 6 | 768 | 151 |
Set Membership: |
Amplicon length greater than or equal to 101 Recommended Assay Caenorhabditis elegans Probe spans exons |