Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gck-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001373560.4 | NP_001360654.1 | 4 | 1277 | 69 | |
NM_077889.4 | NP_510290.1 | 4 - 5 | 1294 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |