Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: twk-23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001270300.3 | NP_001257229.1 | 3 - 4 | 297 | 86 | |
NM_001270301.4 | NP_001257230.1 | 2 - 3 | 201 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |