Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: meg-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_077918.8 | NP_510319.1 | 3 - 4 | 1482 | 54 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |