Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nkat-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_077954.4 | NP_510355.1 | 1 - 2 | 136 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |