Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nkat-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_077954.4 | NP_510355.1 | 3 - 4 | 759 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |