Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sym-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_077960.7 | NP_510361.1 | 6 - 7 | 1302 | 90 | |
GenBank mRNA | AY372076.1 | - | 4 - 5 | 1093 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |