Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ceh-36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_077964.5 | NP_510365.1 | 3 - 4 | 475 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |