Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: del-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001368544.2 | NP_001355359.1 | 6 - 7 | 659 | 81 | |
NM_001368545.3 | NP_001355470.1 | 7 | 803 | 81 | ||
NM_001368546.2 | NP_001355471.1 | 6 - 7 | 608 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |