Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ram-5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_078021.4 | NP_510422.2 | 5 - 6 | 540 | 65 | |
GenBank mRNA | AF218866.1 | - | 5 - 6 | 562 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |