Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nhr-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_078022.7 | NP_510423.1 | 4 - 5 | 845 | 92 | |
GenBank mRNA | AF083222.1 | - | 4 - 5 | 856 | 92 | |
AF273769.1 | - | 4 - 5 | 852 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |